Environmental standard for TOC, BOD, CBOD, COD, DOC demand - whole volume, certified reference material, Supelco®
Catalog # QC3130-500ML
Supplier: Merck
CAS Number:
Specifications
- Matrix:H₂O
- Pk:500 mL
- Description:Demand - Whole Volume
- Environmentally Preferable:
Specifications
About this item
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.