silv-Air 7312 Hygiene Masks
UVEX8707.312
: UVEX
- Type:FFP3 R D preformed mask with valve
- Colour:White
- Pk:5
This FFP3 preformed mask with an exhalation valve, featuring an innovative design and an optimised shape.
- Four-point headband attachment for a comfortable, individual and secure fit
- Extra-wide fabric headband
- Ultrafine porous sealing lip
- Nose clip integrated into the material for comfortable and secure positioning of the mask
The three- dimensional design enables a perfect fit for the many different face shapes which exist.