Polyadenylic acid (poly-A)
Supplier: Merck
Certificates
About this item
Polyadenylic acid (poly-A) is used to evaluate binding on cationic liposomes doped with non-ionic nucleolipids. Poly-A is used in small molecule mRNA targeted drug development to evaluate the binding of potential therapeutic agents such as the isoquinoline group of alkaloids.
Frequently Bought Together