Order Entry
Poland
Orders LinkContactUsLinkComponent
Absorbent mats, SpillEx

Absorbent mats, SpillEx

510×370 mm
Catalog # VILE146760
Supplier:   FHP VILEDA SCS
Absorbent mats, SpillEx
Catalog # VILE146760
Supplier:  FHP VILEDA SCS
Absorbent mats, SpillEx

510×370 mm

Price
246 zł
Log in for availability
1 * 20 SZT
Product Details & Documents

Disposable absorption mats are designed to absorb water-based liquids very quickly.

  • Single use, absorbs up to 1200 ml of water-based liquid
  • Absorbent polymers turn into a gel in contact with liquids
Specifications
  • L×W:
    510×370 mm