Order Entry
Poland
ContactUsLinkComponent
Compound E ≥95% (by HPLC) Calbiochem®, Sigma-Aldrich®
Compound E ≥95% (by HPLC) Calbiochem®, Sigma-Aldrich®
Catalog # 565790-1MG
CAS Number:  
Compound E ≥95% (by HPLC) Calbiochem®, Sigma-Aldrich®
Catalog # 565790-1MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity (by HPLC):
    ≥95%

Specification Test Results

About this item

 :
 :490.511 g/mol
 :Packaged under inert gas.
Storage Temperature:Refrigerator
CAS Number:209986-17-4
Formula:C₂₇H₂₄F₂N₄O₃
MDL Number:MFCD04974531