Order Entry
Poland
ContactUsLinkComponent
 
Dithiothreitol (DTT, Cleland's reagent) ≥99.4%, OmniPur® Calbiochem®, Millipore®
  3870-25GM
 :  
undefined
Dithiothreitol (DTT, Cleland's reagent) ≥99.4%, OmniPur® Calbiochem®, Millipore®
  3870-25GM
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • A (0.02 M in water):
    ≤0.05 AU
  • Assay (-SH content):
    ≥99.4%
  • DNAse:
    None Detected
  • Infrared Spectrum:
    Conforms to Standard
  • Loss on Drying:
    ≤0.5%
  • Melting Point:
    39 - 43 °C
  • Protease:
    None Detected
  • RNAse:
    None Detected

 

 

 :
 :154.25 g/mol
 :GHS07
 :Freezer
 :3483-12-3
 :27565-41-9
 :C₄H₁₀O₂S₂
 :MFCD00004877