Medical keyboards, Induproof® smart compact
Dostawca: GETT GERATETECHNIK
Certyfikaty
Informacje o tym produkcie
Compact silicone keyboard with 86 keys including num pad for use in cramped working environments.
- Ergonomic design
- White housing colour
- Complies with USB 2.0
- Flat key profile for easy cleaning
- Comfortable typing
- IP68 protection, can be submerged into liquids
Keylock function to disable the keys while cleaning the keyboard.
Can be cleaned with soapy water, disinfectants or other cleaning agents.
Certifications: CE, FCC certified.
Powiązane informacje
Frequently Bought Together