Order Entry
Poland
ContactUsLinkComponent
PFTBA (MS tuning compound)
PFTBA (MS tuning compound)
Catalog # REST30482
Supplier:  Restek
CAS Number:  
undefined
PFTBA (MS tuning compound)
Catalog # REST30482
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1 mL
  • Description:
    PFTBA
  • Seq:
    03
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This contains perfluorotributylamine (PFTBA).