Order Entry
Poland
ContactUsLinkComponent
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥96.5% (by HPLC), Sigma-Aldrich®
Irritant
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥96.5% (by HPLC), Sigma-Aldrich®
Catalog # SIAL463310-25G
Supplier:  Merck
CAS Number:  
Irritant
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥96.5% (by HPLC), Sigma-Aldrich®
Catalog # SIAL463310-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Color) :
    White to Faint Yellow to Yellow or Faint Brown to Brown
  • Appearance (Form) :
    Powder
  • Infrared Spectrum:
    Conforms to Structure
  • Morpholine Titration by HCl:
    96.5 - 103.5%
  • Purity (HPLC):
    ≥96.5%

Specification Test Results

About this item

 :
 :294.2200 g/mol
Melting Pt:299...305 °C (lit.)
 :M-Clarity™ quality level = MQ100
EINECS:219-342-9
 :GHS07
CAS Number:2420-87-3
Formula:C₁₆H₆O₆
MDL Number:MFCD00039140