Titanium(IV) oxide 99-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:13463-67-7
:1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:13,09549
Boiling Pt:>2.000 °C
Melting Pt:>1.800 °C
Storage Temperature:Ambient
EINECS:236-675-5
Specification Test Results
- Assay:98.0 - 100.5 %
- Assay (calculated to the dried substance):99.0 - 100.5 %
- Water soluble matter:Max. 0.25 %
- Water soluble matter (hot):Max. 0.5 %
- In acid (HCl 0,5 mol/l) Solution matter:Max. 0.5 %
- Loss on drying (105°C, 3h):Max. 0.5 %
- Loss on ignition (800 °C):Max. 0.5 %
- Acidity or alkalinity:complying
- Arsenic (As):Max. 20 ppm
- Barium (Ba):complying
- Iron (Fe):Max. 50 ppm
- Antimony (Sb):Max. 50 ppm
- Heavy metals (as Pb):Max. 20 ppm
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together