Order Entry
Poland
ContactUsLinkComponent
Mercury(II) acetate ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Toxic
Health
Mercury(II) acetate ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Catalog # 83352-250G
CAS Number:  
Toxic
Health
Mercury(II) acetate ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Catalog # 83352-250G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Insoluble in acetic acid:
    Max. 0.01 %
  • Residue on ignition a. reducing with HCOOH (SO₄)*:
    Max. 0.02 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Mercury (Hg⁺):
    Max. 0.4 %
  • Other heavy metals (as Pb):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.005 %
  • Nitrate (NO₃):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :318.68 g/mol
Merck Index:13,05898
Melting Pt:179-182 °C (dec.)
Melting Pt:179…182 °C (dec.)
Density:3.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-491-1
UN:1629
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:1600-27-7
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165