Order Entry
Poland
ContactUsLinkComponent
Hydrogen peroxide 30% (w/w) in water contains inhibitor ACS
Hydrogen peroxide 30% (w/w) in water contains inhibitor ACS
Certificates
Hydrogen peroxide 30% (w/w) in water contains inhibitor ACS
Certificates

Specification Test Results

  • Assay:
    29.0 - 32.0 %
  • Non-volatile matter:
    Max. 0.002 %
  • Titrable Acid:
    Max. 0.0006 meq/g
  • Iron (Fe):
    Max. 0.5 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Ammonium (NH₄):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 2 ppm
  • Nitrate (NO₃):
    Max. 2 ppm
  • Phosphate (PO₄):
    Max. 2 ppm
  • Sulfate (SO₄):
    Max. 5 ppm
  • APHA:
    Max. 10
  • Vapor Density:
    1 (vs air)

Specification Test Results

Related Information

Frequently Bought Together