Order Entry
Poland
ContactUsLinkComponent
 
 
 
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
    
 
undefined
 
 
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • identity (IR):
    complying
  • assay (GC):
    Min. 99.0 %
  • density (D 20/4):
    0.790 - 0.792
  • density (D 25/25):
    Max. 0.789
  • non-volatile matter:
    Max. 20 ppm
  • water (Karl Fischer):
    Max. 0.3 %
  • acidity or alkalinity:
    complying
  • heavy metals (as Pb):
    Max. 0.0001 %
  • related subst. (GC):
    complying
  • benzene (GC):
    Max. 2 Vol.ppm
  • methanol (GC):
    Max. 0.05 Vol.%
  • 2-propanol (GC):
    Max. 0.05 Vol.%
  • reducing impurities:
    complying
  • water insoluble substances:
    complying
  • residual solvents:
    complying
  • appearance of the solution:
    complying
  • Vapor Pressure:
    184 mmHg ( 20 °C)

 

 

Frequently Bought Together