

Iodine 99.8-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Melting Pt:113.5 °C
Density:4.930 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS08
:GHS07
Specification Test Results
- Vapor Density:9 (vs air)
- Assay:99.8 - 100.5 %
- Non-volatile matter:Max. 0.03 %
- Bromide, chloride (as Cl):Max. 0.01 %
- Residual solvents:complying
- Vapor Pressure:0.31 mmHg ( 25 °C)
Specification Test Results
Frequently Bought Together