Order Entry
Poland
ContactUsLinkComponent
 
Analytical standard, Mecoprop-(4-chloro-2-methylphenoxy-d₆), reference material, Honeywell Fluka™
  HONCR1295-10MG
 :  
Analytical standard, Mecoprop-(4-chloro-2-methylphenoxy-d₆), reference material, Honeywell Fluka™
  HONCR1295-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1705649-54-2
  • Description:
    Mecoprop-(4-chloro-2-methylphenoxy-d₆)
  • Melting point:
    81…85 °C
  • Formula:
    C₁₀D₆H₅ClO₃
  • Storage temperature:
    Ambient
  • Description:
    Mecoprop-(4-chloro-2-methylphenoxy-d₆)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results