Order Entry
Poland
ContactUsLinkComponent
Analytical standard, Cyclosporin A, reference material, Honeywell Fluka™
Analytical standard, Cyclosporin A, reference material, Honeywell Fluka™
  HONCR1136-50MG
 :  
Analytical standard, Cyclosporin A, reference material, Honeywell Fluka™
  HONCR1136-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    59865-13-3
  • Description:
    Cyclosporin A
  • Formula:
    C₆₂H₁₁₁N₁₁O₁₂
  • Storage temperature:
    Ambient
  • Description:
    Cyclosporin A

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results