
Potassium iodide 99.0-100.5% USP, BP, Ph. Eur., pharma grade
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Boiling Pt:1330 °C
Melting Pt:680 °C
EINECS:231-659-4
:GHS08
Specification Test Results
- Assay calc. a.d.s:99.0 - 100.5%
- Alkalinity:Passes Test
- Insoluble matter in H₂O:0.01%
- Loss on drying at 105 °C:1.0%
- Sulfate (SO₄):0.015%
- Residual Solvents (Ph. Eur./USP):Passes Test
- Chloride and Bromide (as Cl):0.05%
- Nitrate, Nitrite and Ammonia:Passes Test
- Thiosulfate:Passes Test
- Iodate:Passes Test
- Heavy metals (as Pb):0.001%
- Ba:Passes Test
- Fe:0.002%
- Cd, Class 1 - (ICH Q3D):0.5 ppm
- Pb, Class 1 - (ICH Q3D):0.5 ppm
- As, Class 1 - (ICH Q3D):1.5 ppm
- Hg, Class 1 - (ICH Q3D):1.5 ppm
- Co, Class 2A - (ICH Q3D):5 ppm
- V, Class 2A - (ICH Q3D):10 ppm
- Ni, Class 2A - (ICH Q3D):20 ppm
- Tl, Class 2B - (ICH Q3D):0.8 ppm
- Au, Class 2B - (ICH Q3D):10 ppm
- Pd, Class 2B - (ICH Q3D):10 ppm
- Ir, Class 2B - (ICH Q3D):10 ppm
- Os, Class 2B - (ICH Q3D):10 ppm
- Rh, Class 2B - (ICH Q3D):10 ppm
- Ru, Class 2B - (ICH Q3D):10 ppm
- Se, Class 2B - (ICH Q3D):15 ppm
- Ag, Class 2B - (ICH Q3D):15 ppm
- Pt, Class 2B - (ICH Q3D):10 ppm
- Li, Class 3 - (ICH Q3D):55 ppm
- Sb, Class 3 - (ICH Q3D):120 ppm
- Ba, Class 3 - (ICH Q3D):140 ppm
- Mo, Class 3 - (ICH Q3D):25 ppm
- Cu, Class 3 - (ICH Q3D):250 ppm
- Sn, Class 3 - (ICH Q3D):600 ppm
- Cr, Class 3 - (ICH Q3D):25 ppm
Specification Test Results
Frequently Bought Together