Iron(III) perchlorate hydrate, yellow crystals, Sigma-Aldrich®
SIAL309281-100G
: Merck
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
- ICP Major Analysis, Confirms Iron Component:Confirmed
- Titration by Na₂S₂O₃:10.0 - 12.5%
- Chloride Content:≤0.10%
:
:372.198 g/mol
:236-908-0
:1481
:GHS03
:GHS07
:15201-61-3
:Cl₃FeH₂O₁₃
:MFCD00149705
:5.1 II
:M-Clarity™ quality level = MQ100