


Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether, M.W. 440 ∼Mn, Sigma-Aldrich®
Catalog # SIAL406708-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Appearance (Color):Colorless to Very Faint Yellow
- Appearance (Form):Liquid
- Infrared Spectrum:Conforms to Structure
- Color Test:< 50 APHA
- Water Content:< 0.25 %
- Total Amines:6.1 - 6.6 meq/g
- Primary Amine:90 - 100 %
Specification Test Results
About this item
:
Density:0.981 g/mL (25 °C) (lit.)
Flash Pt:113 °C
ADR:9,III
:M-Clarity™ quality level = MQ100
UN:3082
:GHS05
:GHS07
:GHS09
CAS Number:39423-51-3
Formula:C₂H₅C[CH₂[OCH₂CH(CH₃)]nNH₂]₃
MDL Number:MFCD00197951