Polyethylene glycol 400 BP, USP, NF, Reag. Ph. Eur., pharma grade
:25322-68-3
:
:H(OCH₂CH₂)nOH
:MFCD00081839
:400 g/mol
:4…8 °C
:1.127 g/cm³
:Ambient
:203-473-3
- Identity According to Pharmacopoeias:Passes Test
- pH of 5% Solution:4.5 - 7.5
- Viscosity Dynamic at 20 °C:105 - 130 mPa
- Viscosity Kinematic at 20 °C:94 - 116 cSt
- Viscosity 98.9 ±0.3 °C:6.8 - 8.0 cSt
- Average Molecular Weight:380 - 420 g/mol
- Acidity or Alkalinity:Passes Test
- Insoluble Matter in H₂O:Passes Test
- Reducing Substances:Passes Test
- Residue on Ignition (as SO₄):Max. 0.1%
- Hydroxyl Value:Max. 264 - 300
- Residual Solvents (Ph. Eur./USP):Passes Test
- Ethylene Oxide (G.C.):Max. 0.0001%
- 1,4-Dioxan (G.C.):Max. 0.001%
- Formaldehyde (CH₂O):Max. 0.0015%
- Ethylene Glycol and Diethylene Glycol (G.C.):Max. 0.25%
- Water (H₂O):Max. 2%
- Heavy metals (as Pb):Max. 0.0005%
- Cd, Class 1 - (ICH Q3D):Max. 0.5 ppm
- Pb, Class 1 - (ICH Q3D):Max. 0.5 ppm
- As, Class 1 - (ICH Q3D):Max. 1.5 ppm
- Hg, Class 1 - (ICH Q3D):Max. 1.5 ppm
- Co, Class 2A - (ICH Q3D):Max. 5 ppm
- V, Class 2A - (ICH Q3D):Max. 10 ppm
- Ni, Class 2A - (ICH Q3D):Max. 20 ppm
- Tl, Class 2B - (ICH Q3D):Max. 5 ppm
- Au, Class 2B - (ICH Q3D):Max. 10 ppm
- Pd, Class 2B - (ICH Q3D):Max. 10 ppm
- Ir, Class 2B - (ICH Q3D):Max. 10 ppm
- Os, Class 2B - (ICH Q3D):Max. 10 ppm
- Rh, Class 2B - (ICH Q3D):Max. 10 ppm
- Ru, Class 2B - (ICH Q3D):Max. 10 ppm
- Se, Class 2B - (ICH Q3D):Max. 15 ppm
- Ag, Class 2B - (ICH Q3D):Max. 15 ppm
- Pt, Class 2B - (ICH Q3D):Max. 10 ppm
- Li, Class 3 - (ICH Q3D):Max. 55 ppm
- Sb, Class 3 - (ICH Q3D):Max. 120 ppm
- Ba, Class 3 - (ICH Q3D):Max. 140 ppm
- Mo, Class 3 - (ICH Q3D):Max. 25 ppm
- Cu, Class 3 - (ICH Q3D):Max. 250 ppm
- Sn, Class 3 - (ICH Q3D):Max. 600 ppm
- Cr, Class 3 - (ICH Q3D):Max. 25 ppm
Frequently Bought Together