Order Entry
Poland
ContactUsLinkComponent
 
Potassium dihydrogen phosphate 98.0-100.5% (by acidimetry) USP, NF, BP, Ph. Eur., pharma grade
 
Potassium dihydrogen phosphate 98.0-100.5% (by acidimetry) USP, NF, BP, Ph. Eur., pharma grade
 

 

  • Assay (Acidimetry):
    98.0-100.5%
  • pH (5% Solution):
    4.2-4.5
  • Loss on drying (130 °C):
    1.0%
  • Insoluble matter in H₂O:
    0.2%
  • Reducing substances:
    Passes test
  • Chloride (Cl):
    0.02%
  • Sulphate (SO₄):
    0.03%
  • Residual solvents (Ph. Eur./USP):
    Passes test
  • Fluoride (F):
    0.001%
  • Heavy metals (as Pb):
    0.001%
  • As:
    0.0001%
  • Fe:
    0.001%
  • PB:
    0.00005%

 

Frequently Bought Together