Order Entry
Poland
ContactUsLinkComponent
Hexadecyltrimethylammonium hydroxide 10% (w/w) in water, Sigma-Aldrich®
Corrosive
Hexadecyltrimethylammonium hydroxide 10% (w/w) in water, Sigma-Aldrich®
Catalog # SIAL439231-25ML
Supplier:  Merck
CAS Number:  
Corrosive
Hexadecyltrimethylammonium hydroxide 10% (w/w) in water, Sigma-Aldrich®
Catalog # SIAL439231-25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :301.56 g/mol
Density:0.988 g/ml (25 °C)
Storage Temperature:Refrigerator
ADR:8,III
 :M-Clarity™ quality level = MQ200
UN:3267
 :GHS05
CAS Number:505-86-2
Formula:C₁₉H₄₃NO
MDL Number:MFCD00041921