


1,3,5-Trioxane for synthesis, Sigma-Aldrich®
: Merck
:110-88-3
:
:C₃H₆O₃
:MFCD00006563
:90.08 g/mol
:12,09863
:114.5 °C (1013 hPa)
:60…62 °C
:1.17 g/cm³ (20 °C)
:203-812-5
:1325
:4.1,II
:M-Clarity™ quality level = MQ200
:38 °C
:GHS02
:GHS08
:GHS07
:Ambient
- Solubility:221 g/l (25 °C)
- Melting Point:60 - 62 °C
- Molar Mass:90.08 g/mol
- Boiling Point:114 - 115 °C (1013 hPa)
- Vapor Pressure:7.5 hPa (20 °C)
- Flash Point:45 °C
- Explosion Limit:3.6 - 28.7 %(V)
- Density:1.17 g/cm3 (65 °C)
- pH:(H2O) neutral
- Ignition Point:410 °C
Frequently Bought Together