Dicyclopentadiene, Sigma-Aldrich®
: Merck
:77-73-6
:
:C₁₀H₁₂
:MFCD00078246
:132.21 g/mol
:13,02767
:170 °C (1013 hPa)
:12…34 °C
:0.982 g/cm³ (20 °C)
:201-052-9
:2048
:3,III
:M-Clarity™ quality level = MQ200
:27 °C
:GHS02
:GHS07
:GHS09
:Ambient
Frequently Bought Together