
Thiosemicarbazide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-19-6
:
Formula:CH₅N₃S
MDL Number:MFCD00007620
:91.14 g/mol
Merck Index:12,09499
Melting Pt:178…180 °C
EINECS:201-184-7
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
Frequently Bought Together