Order Entry
Poland
ContactUsLinkComponent
Octamethyltrisiloxane, 1.0 cSt (25 °C), Sigma-Aldrich®
 
Octamethyltrisiloxane, 1.0 cSt (25 °C), Sigma-Aldrich®
  SIAL469319-50ML
 :  Merck
 :  
undefined
 
Octamethyltrisiloxane, 1.0 cSt (25 °C), Sigma-Aldrich®
  SIAL469319-50ML
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

 

 

 :
 :236.53 g/mol
 :12,06844
 :153 °C (1013 hPa)
 :–82 °C
 :0.82 g/cm³ (20 °C)
 :29 °C (closed cup)
 :203-497-4
 :1993
 :3,III
 :M-Clarity™ quality level = MQ100
 :GHS02
 :107-51-7
 :[(CH₃)₃SiO]₂Si(CH₃)₂
 :MFCD00008264