1,1,3,3,5,5-Hexamethyltrisiloxane, Sigma-Aldrich®
SIAL448621-10ML
: Merck
:
- Pack type:Glass bottle
- Pk:8 mL
- Seq:2265
:
:208.48 g/mol
:128 °C (1013 hPa)
:–67 °C
:0.8246 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:214-716-8
:1189-93-1
:C₆H₂₀O₂Si₃
:MFCD00039788