4-(Dimethylamino)pyridine
: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
- :1122-58-3
- :C₇H₁₀N₂
- :MFCD00006418
- :122.17 g/mol
- :195 °C (1013 hPa)
- :109…111 °C
- :124 °C
- :Ambient
- :214-353-5
- :2811
- :6.1,II
: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
- :1122-58-3
- :C₇H₁₀N₂
- :MFCD00006418
- :122.17 g/mol
- :195 °C (1013 hPa)
- :109…111 °C
- :124 °C
- :Ambient
- :214-353-5
- :2811
- :6.1,II
Product Family Options
- Pack typePk