
2-Chloro-6-methoxy-3-nitropyridine, Sigma-Aldrich®
Catalog # SIALC49909-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:188.57 g/mol
:M-Clarity™ quality level = MQ100
EINECS:253-989-8
:GHS07
CAS Number:38533-61-8
Formula:C₆H₅ClN₂O₃
MDL Number:MFCD00130268