Pyrene, Sigma-Aldrich®
numer katalogowy SIAL571245-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.26 g/mol
Merck Index:12,08147
Boiling Pt:404 °C (1013 hPa)
Melting Pt:148…150 °C
Density:1.27 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:200…220 °C
EINECS:204-927-3
CAS Number:129-00-0
Formula:C₁₆H₁₀
MDL Number:MFCD00004136