
Adenine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:73-24-5
:
Formula:C₅H₅N₅
MDL Number:MFCD00041790
:135.13 g/mol
Merck Index:13,00152
Density:0.99741 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:200-796-1
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS06
Frequently Bought Together