
Adenine
Catalog # MOLEM84081419
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:135.13 g/mol
Merck Index:13,00152
Density:0.99741 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:200-796-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:73-24-5
Formula:C₅H₅N₅
MDL Number:MFCD00041790