Human Recombinant GDNF (from E. coli)
Supplier: Shenandoah Biotechnology
About this item
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
- High quality
- Low endotoxin
- Affordable
: For research use only.
4 Options Available Below
Product Details & Documents
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
- High quality
- Low endotoxin
- Affordable
: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-02-10UG
- SHBT100-02-100UG
- SHBT100-02-500UG
- SHBT100-02-1MG
- Protein/peptide name:
- GDNF
- GDNF
- GDNF
- GDNF
- Protein synonyms:
- ATF-1
- ATF-1
- ATF-1
- ATF-1
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P39905
- P39905
- P39905
- P39905
- Sequence:
- MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
- MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
- MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
- MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mg
- Bioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mg
- Bioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mg
- Bioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0
- 10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0
- 10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0
- 10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0
- Molecular weight:
- 15,2/30,4 kDa
- 15,2/30,4 kDa
- 15,2/30,4 kDa
- 15,2/30,4 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-02-10UGProtein/peptide nameGDNFProtein synonymsATF-1Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP39905SequenceMSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mgPurity≥95%GradeRUOFormulation10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0Molecular weight15,2/30,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-02-100UGProtein/peptide nameGDNFProtein synonymsATF-1Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP39905SequenceMSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mgPurity≥95%GradeRUOFormulation10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0Molecular weight15,2/30,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-02-500UGProtein/peptide nameGDNFProtein synonymsATF-1Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP39905SequenceMSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mgPurity≥95%GradeRUOFormulation10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0Molecular weight15,2/30,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-02-1MGProtein/peptide nameGDNFProtein synonymsATF-1Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP39905SequenceMSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mgPurity≥95%GradeRUOFormulation10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0Molecular weight15,2/30,4 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay



