Tetrahydropyran 99%
:142-68-7
:
:C₅H₁₀O
:MFCD00006585
:88
:–45 °C
:0.88 g/cm³
:86.13 g/mol
:–15 °C
:3271
:3 II
:12,09356
:205-552-8
:GHS07
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4220 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together