
Cobalt(II) hydroxide, Sigma-Aldrich®
Catalog # SIAL342440-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:92.95 g/mol
Merck Index:13,02467
Density:3.597 g/cm³ (15 °C)
:M-Clarity™ quality level = MQ200
EINECS:244-166-4
:GHS07
CAS Number:21041-93-0
Formula:H₂CoO₂
MDL Number:MFCD00016015