
Cobalt(II) hydroxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21041-93-0
:
Formula:H₂CoO₂
MDL Number:MFCD00016015
:92.95 g/mol
Merck Index:14,2442
Melting Pt:250 °C (dec.)
Density:3.597 g/cm³ (15 °C)
EINECS:244-166-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together