
Ruthenium(IV) oxide, anhydrous 99.5+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12036-10-1
:
Formula:O₂Ru
MDL Number:MFCD00074857
Melting Pt:1200 °C
Density:6.97 g/cm³
EINECS:234-840-6
ADR:5.1,II
:133.07 g/mol
:GHS07
Specification Test Results
- Appearance:Black Powder
- Purity:≥99.50 %
Specification Test Results
Frequently Bought Together