Order Entry
Poland
ContactUsLinkComponent
Platinium(IV) oxide 83% Pt
Platinium(IV) oxide 83% Pt
Certificates
undefined
Platinium(IV) oxide 83% Pt

Specification Test Results

  • Appearance:
    Brown Powder
  • Assay:
    ≥79 % (Pt)

Specification Test Results

Related Information

Frequently Bought Together