

Os EnCat® 40, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:20816-12-0
:
Formula:O₄Os
MDL Number:MFCD00011150
:254.23 g/mol
Merck Index:13,06962
Boiling Pt:130 °C (1013 hPa)
Melting Pt:40 °C
Density:1.04 g/cm³ (20 °C)
UN:2471
ADR:6.1,I
:M-Clarity™ quality level = MQ100
EINECS:244-058-7
:GHS06
:GHS05
Storage Temperature:Ambient
Frequently Bought Together