Order Entry
Poland
ContactUsLinkComponent
Iron(III) oxide, powder, NanoArc® 20-40 nm, magnetic S.A. 30-60m2/g
Iron(III) oxide, powder, NanoArc® 20-40 nm, magnetic S.A. 30-60m2/g
Catalog # 44896.36
CAS Number:  
Iron(III) oxide, powder, NanoArc® 20-40 nm, magnetic S.A. 30-60m2/g
Catalog # 44896.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :159.69 g/mol
Merck Index:13,04058
Melting Pt:1565 °C
Density:5.24
EINECS:215-168-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1309-37-1
Formula:Fe₂O₃
MDL Number:MFCD00011008