Iron(III) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1309-37-1
:
Formula:Fe₂O₃
MDL Number:MFCD00011008
:159.69 g/mol
Merck Index:14,4028
Melting Pt:1565 °C
Density:5.24
EINECS:215-168-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Total Metal Impurities:0.002% max
Specification Test Results
Frequently Bought Together