
Iron(II,III) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1317-61-9
:
Formula:Fe₃O₄
MDL Number:MFCD00011010
:231.54 g/mol
Merck Index:12,04087
Melting Pt:1560 °C
Density:5.18 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:215-277-5
:GHS07
Frequently Bought Together