

Cobalt(II,III) oxide ≥99% (metals basis), nanopowder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1308-06-1
:
Formula:Co₃O₄
MDL Number:MFCD00010939
:240.8 g/mol
Merck Index:14,2429
Melting Pt:900 °C (dec.)
Density:6.11
Storage Temperature:Ambient
EINECS:215-157-2
UN:3077
ADR:9,III
:GHS08
:GHS07
Related Information
Frequently Bought Together