Order Entry
Poland
ContactUsLinkComponent
Palladium(II) chloride (59% Pd) hydrogenation catalyst
Irritant
Palladium(II) chloride (59% Pd) hydrogenation catalyst
Catalog # 26136.083
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Palladium(II) chloride (59% Pd) hydrogenation catalyst
Catalog # 26136.083
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Assay (on dried substance):
    Min. 99 %

Specification Test Results

About this item

 :
Merck Index:13,07058
Melting Pt:678 °C
Density:4 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,III
 :177.33 g/mol
EINECS:231-596-2
UN:3260
 :GHS07
CAS Number:7647-10-1
Formula:PdCl₂
MDL Number:MFCD00003558