

Manganese(II) chloride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7773-01-5
:
Formula:MnCl₂
MDL Number:MFCD00011114
:125.84 g/mol
Merck Index:14,5728
Boiling Pt:1190 °C
Melting Pt:650 °C
Density:2.977
UN:3288
ADR:6.1,III
:ampouled under argon
EINECS:231-869-6
:GHS06
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together