



Manganese(II) chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7773-01-5
:
Formula:MnCl₂
MDL Number:MFCD00011114
:125.84 g/mol
Merck Index:12,05768
Boiling Pt:1190 °C (760 mmHg)
Melting Pt:650 °C
Density:2,98 g/cm³ (20 °C)
EINECS:231-869-6
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Pink Flakes or beads
- Titration Complexometric:≥96.0 %
- Water:≤1 %
- Iron (Fe):≤200 ppm
- Nickel (Ni):≤200 ppm
Specification Test Results
Frequently Bought Together