Order Entry
Poland
Orders LinkContactUsLinkComponent
Human Recombinant EBI3 (from <i>E. coli</i>)

Human Recombinant EBI3 (from E. coli)

Human Recombinant EBI3 (from <i>E. coli</i>)
Certificates
Human Recombinant EBI3 (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Epstein-Barr virus induced gene-3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-162-5UG
  • SHBT100-162-20UG
  • SHBT100-162-100UG
  • SHBT100-162-500UG
  • SHBT100-162-1MG
  • Protein/peptide name:
  • EBI3 subunit (IL-27/IL-35)
  • EBI3 subunit (IL-27/IL-35)
  • EBI3 subunit (IL-27/IL-35)
  • EBI3 subunit (IL-27/IL-35)
  • EBI3 subunit (IL-27/IL-35)
  • Protein synonyms:
  • IL-35/EBI3|IL-27/EBI3
  • IL-35/EBI3|IL-27/EBI3
  • IL-35/EBI3|IL-27/EBI3
  • IL-35/EBI3|IL-27/EBI3
  • IL-35/EBI3|IL-27/EBI3
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q14213
  • Q14213
  • Q14213
  • Q14213
  • Q14213
  • Sequence:
  • MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
  • MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
  • MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
  • MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
  • MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤5
  • Endotoxin LAL (EU/µg) ≤5
  • Endotoxin LAL (EU/µg) ≤5
  • Endotoxin LAL (EU/µg) ≤5
  • Endotoxin LAL (EU/µg) ≤5
  • UniProtKB:
  • Q14213
  • Q14213
  • Q14213
  • Q14213
  • Q14213
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥90%
  • ≥90%
  • ≥90%
  • ≥90%
  • ≥90%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
  • 0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
  • 0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
  • 0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
  • 0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
  • Molecular weight:
  • 23,4 kDa
  • 23,4 kDa
  • 23,4 kDa
  • 23,4 kDa
  • 23,4 kDa
  • Reconstitution instructions:
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG - 326 zł
    326 zł
    1 * 5 µG
       326 zł

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-162-5UG
    Protein/peptide name
    EBI3 subunit (IL-27/IL-35)
    Protein synonyms
    IL-35/EBI3|IL-27/EBI3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q14213
    Sequence
    MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤5
    UniProtKB
    Q14213
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
    Molecular weight
    23,4 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG - 809 zł
    809 zł
    1 * 20 µG
       809 zł

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-162-20UG
    Protein/peptide name
    EBI3 subunit (IL-27/IL-35)
    Protein synonyms
    IL-35/EBI3|IL-27/EBI3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q14213
    Sequence
    MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤5
    UniProtKB
    Q14213
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
    Molecular weight
    23,4 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 3 130 zł
    3 130 zł
    1 * 100 µG
       3 130 zł

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-162-100UG
    Protein/peptide name
    EBI3 subunit (IL-27/IL-35)
    Protein synonyms
    IL-35/EBI3|IL-27/EBI3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q14213
    Sequence
    MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤5
    UniProtKB
    Q14213
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
    Molecular weight
    23,4 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 15 540 zł
    15 540 zł
    1 * 500 µG
       15 540 zł

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-162-500UG
    Protein/peptide name
    EBI3 subunit (IL-27/IL-35)
    Protein synonyms
    IL-35/EBI3|IL-27/EBI3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q14213
    Sequence
    MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤5
    UniProtKB
    Q14213
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
    Molecular weight
    23,4 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 25 910 zł
    25 910 zł
    1 * 1 mg
       25 910 zł

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-162-1MG
    Protein/peptide name
    EBI3 subunit (IL-27/IL-35)
    Protein synonyms
    IL-35/EBI3|IL-27/EBI3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q14213
    Sequence
    MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤5
    UniProtKB
    Q14213
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
    Molecular weight
    23,4 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT