Niobium(IV) oxide, Sigma-Aldrich®
Catalog # SIAL383163-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:124.91 g/mol
Melting Pt:1915 °C
Density:5.98 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:234-809-7
CAS Number:12034-59-2
Formula:NbO₂
MDL Number:MFCD00016242