Titanium(IV) isopropoxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:546-68-9
:
Formula:C₁₂H₂₈O₄Ti
MDL Number:MFCD00008871
Merck Index:12,09614
Boiling Pt:240 °C (1013 hPa)
Melting Pt:18…20 °C
Density:0.9711 g/cm³ (20 °C)
Flash Pt:51 °C
EINECS:208-909-6
UN:2413
ADR:3,III
:M-Clarity™ quality level = MQ200
:284.23 g/mol
Frequently Bought Together