

Titanium(IV) isopropoxide 98+%
:546-68-9
:
:C₁₂H₂₈O₄Ti
:MFCD00008871
:12,09614
:240 °C (760 mmHg)
:>15 °C
:0.95 g/cm³
:46 °C
:2413
:3,III
:284.23 g/mol
:208-909-6
:GHS06
:GHS02
:Ambient
- Appearance:Clear liquid (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:≥15 °C
- Assay:16.6 to 17.3 % (Ti)
- Color scale:≤100 APHA
Frequently Bought Together